accessibility menu, dialog, popup

UserName

CAS RN 887267-47-2

OFFFH2NBr

CAS RN 887267-47-2

IUPAC Name: 2-bromo-5-(trifluoromethoxy)aniline
Synonyms: 2-bromo-5-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 256.02
Molecular Formula: C7H5BrF3NO
InChi Key: MMRYEBVMIOYMIF-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=CC(OC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results