missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-(trifluoromethoxy)aniline 98.0+%, TCI America™

Catalog No. B310725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B310725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B310725G Supplier TCI America Supplier No. B310725G
Only null left

Specifications

CAS 887267-47-2
Color Yellow
Molecular Formula C7H5BrF3NO
Quantity 25 g
MDL Number MFCD08458013
Synonym 2-Amino-1-bromo-4-(trifluoromethoxy)benzene
InChI Key MMRYEBVMIOYMIF-UHFFFAOYSA-N
SMILES NC1=C(Br)C=CC(OC(F)(F)F)=C1
IUPAC Name 2-bromo-5-(trifluoromethoxy)aniline
Molecular Weight (g/mol) 256.02
PubChem CID 26985111
Formula Weight 256.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-(trifluoromethoxy)aniline
Show More Show Less