accessibility menu, dialog, popup

UserName

CAS RN 887144-97-0

IUPAC Name: 3,3-dimethyl-1-(trifluoromethyl)-3H-1λ³,2-benziodaoxole
Synonyms: 3,3-dimethyl-1-(trifluoromethyl)-1λ³,2-benziodaoxole
Molecular Weight (g/mol): 330.09
Molecular Formula: C10H10F3IO
InChi Key: HVAPLSNCVYXFDQ-UHFFFAOYSA-N
SMILES: CC1(C)O[I](C2=CC=CC=C12)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)