CAS RN 88671-89-0
IUPAC Name:
2-(4-chlorophenyl)-2-[(1H-1,2,4-triazol-1-yl)methyl]hexanenitrile
Synonyms:
myclobutanil
Molecular Weight (g/mol):
288.78
Molecular Formula:
C15H17ClN4
InChi Key:
HZJKXKUJVSEEFU-UHFFFAOYNA-N
SMILES:
CCCCC(CN1C=NC=N1)(C#N)C1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals Myclobutanil
Thermo Scientific Chemicals Myclobutanil
CAS: 88671-89-0 Molecular Formula: C15H17ClN4 Molecular Weight (g/mol): 288.78 InChI Key: HZJKXKUJVSEEFU-UHFFFAOYNA-N IUPAC Name: 2-(4-chlorophenyl)-2-[(1H-1,2,4-triazol-1-yl)methyl]hexanenitrile SMILES: CCCCC(CN1C=NC=N1)(C#N)C1=CC=C(Cl)C=C1