missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Myclobutanil

Catalog No. AC467142500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467142500 250 mg
AC467140010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467142500 Supplier Thermo Scientific Chemicals Supplier No. 467142500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 88671-89-0 | C15H17ClN4 | 288.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88671-89-0
Molecular Formula C15H17ClN4
Molecular Weight (g/mol) 288.78
InChI Key HZJKXKUJVSEEFU-UHFFFAOYNA-N
IUPAC Name 2-(4-chlorophenyl)-2-[(1H-1,2,4-triazol-1-yl)methyl]hexanenitrile
SMILES CCCCC(CN1C=NC=N1)(C#N)C1=CC=C(Cl)C=C1

Specifications

Melting Point 63.0°C to 68.0°C
Quantity 250 mg
Formula Weight 288.78
Chemical Name or Material Myclobutanil
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Causes serious eye irritation.

Suspected of damaging the unborn child.

Toxic to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only