accessibility menu, dialog, popup

UserName

H3COOH2NCH3SOHOO

CAS RN 886-86-2

IUPAC Name: ethyl 3-aminobenzoate; methanesulfonic acid
Synonyms: tricaine
Molecular Weight (g/mol): 261.29
Molecular Formula: C10H15NO5S
InChi Key: FQZJYWMRQDKBQN-UHFFFAOYSA-N
SMILES: CS(O)(=O)=O.CCOC(=O)C1=CC=CC(N)=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)