accessibility menu, dialog, popup

UserName

O

CAS RN 886-38-4

IUPAC Name: diphenylcycloprop-2-en-1-one
Synonyms: diphenylcyclopropenone DPCP DPC 2,3-diphenylcyclopropenone diphencyprone
Molecular Weight (g/mol): 206.24
Molecular Formula: C15H10O
InChi Key: HCIBTBXNLVOFER-UHFFFAOYSA-N
SMILES: O=C1C(=C1C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)