CAS RN 885950-21-0
IUPAC Name:
3-bromoimidazo[1,2-a]pyridine-6-carbonitrile
Synonyms:
3-bromoimidazo[1,2-a]pyridine-6-carbonitrile
Molecular Weight (g/mol):
222.05
Molecular Formula:
C8H4BrN3
InChi Key:
UOQHWNPVNXSDDO-UHFFFAOYSA-N
SMILES:
BrC1=CN=C2C=CC(=CN12)C#N
1
–
1
of
1
results
1
Promotions Available
3-Bromo-6-cyanoimidazo[1,2-a]pyridine, 95%
3-Bromo-6-cyanoimidazo[1,2-a]pyridine, 95%
CAS: 885950-21-0 Molecular Formula: C8H4BrN3 Molecular Weight (g/mol): 222.05 MDL Number: MFCD06659617 InChI Key: UOQHWNPVNXSDDO-UHFFFAOYSA-N Synonym: 3-bromoimidazo 1,2-a pyridine-6-carbonitrile,3-bromo-6-cyanoimidazo 1,2-a pyridine,3-bromo-4-hydroimidazo 1,2-a pyridine-6-carbonitrile PubChem CID: 2764571 IUPAC Name: 3-bromoimidazo[1,2-a]pyridine-6-carbonitrile SMILES: BrC1=CN=C2C=CC(=CN12)C#N