missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-6-cyanoimidazo[1,2-a]pyridine, 95%

Catalog No. AAH35421MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH35421MD 250 mg
AAH3542103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH35421MD Supplier Thermo Scientific Chemicals Supplier No. H35421MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 885950-21-0 | C8H4BrN3 | 222.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 885950-21-0
Molecular Formula C8H4BrN3
Molecular Weight (g/mol) 222.05
MDL Number MFCD06659617
InChI Key UOQHWNPVNXSDDO-UHFFFAOYSA-N
Synonym 3-bromoimidazo 1,2-a pyridine-6-carbonitrile, 3-bromo-6-cyanoimidazo 1,2-a pyridine, 3-bromo-4-hydroimidazo 1,2-a pyridine-6-carbonitrile
PubChem CID 2764571
IUPAC Name 3-bromoimidazo[1,2-a]pyridine-6-carbonitrile
SMILES BrC1=CN=C2C=CC(=CN12)C#N

Specifications

Melting Point 207°C to 209°C
Quantity 250 mg
Formula Weight 222.05
Percent Purity 95%
Chemical Name or Material 3-Bromo-6-cyanoimidazo[1,2-a]pyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only