accessibility menu, dialog, popup

UserName

CAS RN 884497-46-5

IUPAC Name: methyl 3-amino-5-fluorobenzoate
Synonyms: methyl 3-amino-5-fluorobenzoate
Molecular Weight (g/mol): 169.16
Molecular Formula: C8H8FNO2
InChi Key: YKEJVXMOMKWEPV-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(N)=CC(F)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results