missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-5-fluorobenzoate, 98%

Catalog No. AAH63038MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63038MD 250 mg
AAH6303803 1 g
AAH6303806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63038MD Supplier Thermo Scientific Chemicals Supplier No. H63038MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 884497-46-5 | C8H8FNO2 | 169.155 g/mol

Methyl 3-amino-5-fluorobenzoate is an amino acid widely used in pharmaceuticals and foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-amino-5-fluorobenzoate is an amino acid widely used in pharmaceuticals and foods.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 884497-46-5
Molecular Formula C8H8FNO2
Molecular Weight (g/mol) 169.155
MDL Number MFCD05863683
InChI Key YKEJVXMOMKWEPV-UHFFFAOYSA-N
Synonym methyl3-amino-5-fluorobenzoate, methyl 5-amino-3-fluorobenzoate, 5-amino-3-fluorobenzoic acid methyl ester, 3-amino-5-fluoro-benzoic acid methyl ester
PubChem CID 3807862
IUPAC Name methyl 3-amino-5-fluorobenzoate
SMILES COC(=O)C1=CC(=CC(=C1)F)N

Specifications

Color White to Yellow
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 169.16
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 3-amino-5-fluorobenzoate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only