CAS RN 88332-30-3
IUPAC Name:
6-methyl-3-(3-methylbutyl)heptan-2-yl tetradecanoate
Synonyms:
isotridecyl myristate
Molecular Weight (g/mol):
410.73
Molecular Formula:
C27H54O2
InChi Key:
JHAGSLRTFDBCLA-UHFFFAOYNA-N
SMILES:
CCCCCCCCCCCCCC(=O)OC(C)C(CCC(C)C)CCC(C)C
1
–
1
of
1
results
1
3-Isoamyl-6-methyl-2-heptyl Myristate 95.0+%, TCI America™
CAS: 88332-30-3 Molecular Formula: C27H54O2 Molecular Weight (g/mol): 410.727 InChI Key: JHAGSLRTFDBCLA-UHFFFAOYSA-N Synonym: 3-Isoamyl-6-methyl-2-heptyl Tetradecanoate, 2-Isopentyl-1,5-dimethylhexyl Tetradecanoate, Myristic Acid 3-Isoamyl-6-methyl-2-heptyl Ester, Tetradecanoic Acid 3-Isoamyl-6-methyl-2-heptyl Ester PubChem CID: 44629799 IUPAC Name: [6-methyl-3-(3-methylbutyl)heptan-2-yl] tetradecanoate SMILES: CCCCCCCCCCCCCC(=O)OC(C)C(CCC(C)C)CCC(C)C