missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Isoamyl-6-methyl-2-heptyl Myristate 95.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Quantity | 500 g |
| Formula Weight | 410.73 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Isoamyl-6-methyl-2-heptyl Myristate |
Safety and Handling
| EINECSNumber | (2)-0798&(2)-2489 |
| TSCA | No |