accessibility menu, dialog, popup

UserName

H3CNNOO

CAS RN 881-07-2

IUPAC Name: 2-methyl-8-nitroquinoline
Synonyms: quinoline, 2-methyl-8-nitro-
Molecular Weight (g/mol): 188.19
Molecular Formula: C10H8N2O2
InChi Key: UHPGVDHXHDPYQP-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=CC=C(C2=N1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)