missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-8-nitroquinoline 98.0+%, TCI America™

Catalog No. N05395G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05395G 5 g
N053925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N05395G Supplier TCI America Supplier No. N05395G
Only null left

Chemical Identifiers

CAS 881-07-2
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.19
MDL Number MFCD00006764
InChI Key UHPGVDHXHDPYQP-UHFFFAOYSA-N
PubChem CID 13433
IUPAC Name 2-methyl-8-nitroquinoline
SMILES CC1=CC=C2C=CC=C(C2=N1)[N+]([O-])=O

Specifications

Melting Point 138°C
Color Yellow
Quantity 5 g
Synonym 8-nitroquinaldine, quinaldine, 8-nitro, quinoline, 2-methyl-8-nitro, unii-xrv949auqz, xrv949auqz, acmc-209qr6, 2-methyl-8-nitro-quinoline, 5-20-07-00387 beilstein handbook reference, quinoline,2-methyl-8-nitro, uhpgvdhxhdpyqp-uhfffaoysa
Formula Weight 188.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-8-nitroquinoline
RTECSNumber UZ9830000
TSCA No