accessibility menu, dialog, popup

UserName

CAS RN 88-67-5

IUPAC Name: 2-iodobenzoic acid
Synonyms: 2-iodobenzoic acid 2-jod-benzoesaeure o-jod-benzoesaeure
Molecular Weight (g/mol): 248.02
Molecular Formula: C7H5IO2
InChi Key: CJNZAXGUTKBIHP-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1I
Molecular Weight (g/mol) 
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)