missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobenzoic Acid 98.0+%, TCI America™

Catalog No. I0053500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0053500G 500 g
I005325G 25 g
I0053100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0053500G Supplier TCI America Supplier No. I0053500G

Chemical Identifiers

CAS 88-67-5
Molecular Formula C7H5IO2
Molecular Weight (g/mol) 248.019
MDL Number MFCD00002419
InChI Key CJNZAXGUTKBIHP-UHFFFAOYSA-N
Synonym o-iodobenzoic acid, benzoic acid, 2-iodo, iodobenzoic acid, benzoic acid, o-iodo, usaf ek-572, 2-iodo benzoic acid, 2-iodo-benzoic acid, benzoic acid, iodo, kyselina o-jodbenzoova, o-jod-benzoesaeure
PubChem CID 6941
ChEBI CHEBI:287979
IUPAC Name 2-iodobenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)I

Specifications

Melting Point 162°C
Quantity 500 g
Formula Weight 248.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodobenzoic Acid
EINECSNumber (3)-3578
RTECSNumber DH2975000
TSCA Yes