accessibility menu, dialog, popup

UserName

CH3ONHH2NSOHOO

CAS RN 88-64-2

IUPAC Name: 2-amino-4-acetamidobenzene-1-sulfonic acid
Synonyms: 2-amino-4-acetamidobenzenesulfonic acid
Molecular Weight (g/mol): 230.24
Molecular Formula: C8H10N2O4S
InChi Key: FOINSAWEWXUXPQ-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C(N)=C1)S(O)(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results