missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamido-2-aminobenzenesulfonic Acid Hydrate 98.0+%, TCI America™

Catalog No. A194425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A194425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A194425G Supplier TCI America Supplier No. A194425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 88-64-2
Molecular Formula C8H10N2O4S
Molecular Weight (g/mol) 230.238
MDL Number MFCD00035752
InChI Key FOINSAWEWXUXPQ-UHFFFAOYSA-N
Synonym 3′C-Amino-4′C-sulfoacetanilide
PubChem CID 6939
IUPAC Name 4-acetamido-2-aminobenzenesulfonic acid
SMILES CC(=O)NC1=CC(=C(C=C1)S(=O)(=O)O)N

Specifications

Quantity 25 g
Formula Weight 230.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetamido-2-aminobenzenesulfonic Acid Hydrate
EINECSNumber (3)-3144
RTECSNumber AE7050000
TSCA Yes