accessibility menu, dialog, popup

UserName

CH3CH3ClSClOO

CAS RN 88-49-3

IUPAC Name: 4-chloro-2,5-dimethylbenzene-1-sulfonyl chloride
Synonyms: 4-chloro-2,5-dimethylbenzenesulfonyl chloride
Molecular Weight (g/mol): 239.11
Molecular Formula: C8H8Cl2O2S
InChi Key: JBYZPUBAISWVDI-UHFFFAOYSA-N
SMILES: CC1=CC(=C(C)C=C1Cl)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results