missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2,5-dimethylbenzenesulfonyl chloride, 98%

Catalog No. AAL0524714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0524714 25 g
AAL0524706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0524714 Supplier Thermo Scientific Chemicals Supplier No. L0524714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 88-49-3 | C8H8Cl2O2S | 239.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88-49-3
Molecular Formula C8H8Cl2O2S
Molecular Weight (g/mol) 239.11
MDL Number MFCD00044017
InChI Key JBYZPUBAISWVDI-UHFFFAOYSA-N
Synonym 5-chloro-p-xylene-2-sulphonyl chloride, 4-chloro-2,5-dimethylbenzene-1-sulfonyl chloride, 4-chloro-2, 5-dimethylphenylsulfonyl chloride, 2,5-dimethyl-4-chlorobenzenesulfonyl chloride, chloro 4-chloro-2,5-dimethylphenyl sulfone, pubchem5091, 4-chloro-2,5-dimethylbenzenesulfonylchloride, 4-chloro 2,5-dimethylphenylsulfonylchloride, 4-chloro-2,5-dimethylphenylsulfonyl chloride, 4-chloro-2,5-dimethyl-benzenesulfonyl chloride
PubChem CID 66618
IUPAC Name 4-chloro-2,5-dimethylbenzenesulfonyl chloride
SMILES CC1=CC(=C(C)C=C1Cl)S(Cl)(=O)=O

Specifications

Melting Point 48°C to 50°C
Quantity 25 g
UN Number UN3261
Beilstein 2838165
Formula Weight 239.12
Percent Purity 98%
Chemical Name or Material 4-Chloro-2,5-dimethylbenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 201-835-5
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only