accessibility menu, dialog, popup

UserName

NH2ClHOSHOOO

CAS RN 88-23-3

IUPAC Name: 3-amino-5-chloro-2-hydroxybenzene-1-sulfonic acid
Synonyms: 3-amino-5-chloro-2-hydroxybenzenesulfonic acid
Molecular Weight (g/mol): 223.63
Molecular Formula: C6H6ClNO4S
InChi Key: YCTAOQGPWNTYJE-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=CC(=C1O)S(O)(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results