missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chlorophenol-6-sulfonic Acid 99.0+%, TCI America™

Catalog No. A077325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A077325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A077325G Supplier TCI America Supplier No. A077325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 88-23-3
Molecular Formula C6H6ClNO4S
Molecular Weight (g/mol) 223.627
MDL Number MFCD00035768
InChI Key YCTAOQGPWNTYJE-UHFFFAOYSA-N
Synonym 3-Amino-5-chloro-2-hydroxybenzenesulfonic Acid
PubChem CID 6927
IUPAC Name 3-amino-5-chloro-2-hydroxybenzenesulfonic acid
SMILES C1=C(C=C(C(=C1S(=O)(=O)O)O)N)Cl

Specifications

Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 223.63
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-chlorophenol-6-sulfonic Acid
EINECSNumber (3)-2090
TSCA Yes