accessibility menu, dialog, popup

UserName

CAS RN 88-17-5

IUPAC Name: 2-(trifluoromethyl)aniline
Synonyms: 2-(trifluoromethyl)aniline
Molecular Weight (g/mol): 161.13
Molecular Formula: C7H6F3N
InChi Key: VBLXCTYLWZJBKA-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1C(F)(F)F
Boiling Point 
  • (4)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (4)
  • (2)
  • (1)
  • (3)
  • (4)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)