missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzotrifluoride, 99%

Catalog No. AC103185000
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103185000 500 mL Glass bottle
AC103180050 5 mL Glass bottle
AC103181000 100 mL Glass bottle
AC103180025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC103185000 Supplier Thermo Scientific Chemicals Supplier No. 103185000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 88-17-5 | C7H6F3N | 161.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88-17-5
Molecular Formula C7H6F3N
Molecular Weight (g/mol) 161.13
MDL Number MFCD00007718
InChI Key VBLXCTYLWZJBKA-UHFFFAOYSA-N
Synonym 2-trifluoromethyl aniline, 2-aminobenzotrifluoride, o-aminobenzotrifluoride, o-trifluoromethyl aniline, o-trifluoromethylaniline, benzenamine, 2-trifluoromethyl, 2-trifluoromethyl benzenamine, alpha,alpha,alpha-trifluoro-o-toluidine, 2-aminotrifluorotoluene, 2-aminotrifluoromethylbenzene
PubChem CID 6922
IUPAC Name 2-(trifluoromethyl)aniline
SMILES C1=CC=C(C(=C1)C(F)(F)F)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -33.0°C
Color Amber to Colorless
Density 1.2960g/mL
Boiling Point 170.0°C to 173.0°C (15.0 mmHg)
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4800 to 1.4820
Linear Formula H2NC6H4CF3
Quantity 500 mL
Beilstein 12, II, 453
Specific Gravity 1.296
Solubility Information Solubility in water: 4g/L in water (20°C). Other solubilities: soluble in ether,ethanol and diethyl ether
Formula Weight 161.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Aminobenzotrifluoride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic lif
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plen
EINECSNumber 201-806-7
TSCA TSCA

RUO – Research Use Only