accessibility menu, dialog, popup

UserName

CAS RN 88-13-1

IUPAC Name: thiophene-3-carboxylic acid
Synonyms: thiophene-3-carboxylic acid
Molecular Weight (g/mol): 128.15
Molecular Formula: C5H4O2S
InChi Key: YNVOMSDITJMNET-UHFFFAOYSA-N
SMILES: OC(=O)C1=CSC=C1
Molecular Weight (g/mol) 
  • (8)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (8)
  • (8)
  • (2)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (4)