missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-3-carboxylic acid, 97%, Thermo Scientific™

Catalog No. p-2704682
Change view
Click to view available options
Quantity:
1g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC13101DA 1g
CC13101EA 10g
CC13101EE 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC13101DA Supplier Thermo Scientific Chemicals Supplier No. CC13101DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 88-13-1
Molecular Formula C5H4O2S
Molecular Weight (g/mol) 128.145
MDL Number MFCD00005467
InChI Key YNVOMSDITJMNET-UHFFFAOYSA-N
Synonym 3-thiophenecarboxylic acid, 3-thenoic acid, 3-thiophenezoic acid, beta-thiophenic acid, 3-thienylcarboxylic acid, 3-thiophene carboxylic acid, 3-carboxythiophene, beta-thiophenecarboxylic acid, 3-thiophenoic acid, unii-6v3012q6be
PubChem CID 6918
IUPAC Name thiophene-3-carboxylic acid
SMILES C1=CSC=C1C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 128.15
Percent Purity 97%
Chemical Name or Material 3-Thiophenecarboxylic acid
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.