accessibility menu, dialog, popup

UserName

CH3OBrOCH3

CAS RN 877-88-3

IUPAC Name: 1-(bromomethyl)-3,5-dimethoxybenzene
Synonyms: 1-(bromomethyl)-3,5-dimethoxybenzene
Molecular Weight (g/mol): 231.09
Molecular Formula: C9H11BrO2
InChi Key: BTHIGJGJAPYFSJ-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=CC(CBr)=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)