missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxybenzyl Bromide 97.0+%, TCI America™

Catalog No. D26575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26575G 5 g
D265725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26575G Supplier TCI America Supplier No. D26575G
Only null left

Chemical Identifiers

CAS 877-88-3
Molecular Formula C9H11BrO2
Molecular Weight (g/mol) 231.089
MDL Number MFCD01321368
InChI Key BTHIGJGJAPYFSJ-UHFFFAOYSA-N
Synonym 3,5-dimethoxybenzyl bromide, 1-bromomethyl-3,5-dimethoxybenzene, 3,5-dimethoxybenzylbromide, benzene, 1-bromomethyl-3,5-dimethoxy, 3,5-dimethoxy benzyl bromide, alpha-bromo-3,5-dimethoxytoluene, 5-bromomethyl-1,3-dimethoxybenzene, 5-bromomethyl resorcinol dimethyl ether, pubchem13588, acmc-209qq1
PubChem CID 1274490
IUPAC Name 1-(bromomethyl)-3,5-dimethoxybenzene
SMILES COC1=CC(=CC(=C1)CBr)OC

Specifications

Melting Point 73°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 231.09
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethoxybenzyl Bromide
TSCA No
Recommended Storage Refrigerator