CAS RN 87691-87-0
CAS RN 87691-87-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-(1-Piperazinyl)-1,2-benzisothiazole 98.0+%, TCI America™
CAS: 87691-87-0 Molecular Formula: C11H13N3S Molecular Weight (g/mol): 219.306 MDL Number: MFCD04117970 InChI Key: KRDOFMHJLWKXIU-UHFFFAOYSA-N PubChem CID: 2772144 IUPAC Name: 3-piperazin-1-yl-1,2-benzothiazole SMILES: C1CN(CCN1)C2=NSC3=CC=CC=C32