accessibility menu, dialog, popup

UserName

CAS RN 875-79-6

IUPAC Name: 1,2-dimethyl-1H-indole
Synonyms: 1,2-dimethylindole
Molecular Weight (g/mol): 145.21
Molecular Formula: C10H11N
InChi Key: BJMUOUXGBFNLSN-UHFFFAOYSA-N
SMILES: CN1C(C)=CC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results