accessibility menu, dialog, popup

UserName

H3CNBr

CAS RN 874-80-6

IUPAC Name: 1-butylpyridin-1-ium bromide
Synonyms: butylpyridinium bromide
Molecular Weight (g/mol): 216.12
Molecular Formula: C9H14BrN
InChi Key: KVBQNFMTEUEOCD-UHFFFAOYSA-M
SMILES: [Br-].CCCC[N+]1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)