missing translation for 'onlineSavingsMsg'
Learn More

1-Butylpyridinium Bromide 98.0+%, TCI America™

Catalog No. B17435G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17435G 5 g
B174325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17435G Supplier TCI America Supplier No. B17435G

Chemical Identifiers

CAS 874-80-6
Molecular Formula C9H14BrN
Molecular Weight (g/mol) 216.12
MDL Number MFCD00671496
InChI Key KVBQNFMTEUEOCD-UHFFFAOYSA-M
Synonym 1-butylpyridinium bromide, butylpyridinium bromide, 1-butylpyridin-1-ium bromide, n-butylpyridinium bromide, unii-f72fd1bswg, f72fd1bswg, 1-n-butylpyridinium bromide, pyridinium, 1-butyl-, bromide, 1-butylpyridiniumbromide, acmc-209qnu
PubChem CID 2734238
IUPAC Name 1-butylpyridin-1-ium bromide
SMILES [Br-].CCCC[N+]1=CC=CC=C1

Specifications

Melting Point 104°C
Color White-Yellow
Quantity 5 g
Formula Weight 216.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Butylpyridinium Bromide