accessibility menu, dialog, popup

UserName

CH3CH3H3CCH3NNCCH3H3CCH3CH3ClAg

CAS RN 873297-19-9

IUPAC Name: 1,3-bis[2,6-bis(propan-2-yl)phenyl]-2,3-dihydro-1H-imidazol-2-ylidene; chlorosilver
Synonyms: 1,3-bis(2,6-diisopropylphenyl)-2H-imidazol-2-ylidene; silver chloride
Molecular Weight (g/mol): 531.92
Molecular Formula: C27H36AgClN2
InChi Key: GNVDTUAJUVJOFQ-UHFFFAOYSA-M
SMILES: Cl[Ag].CC(C)C1=CC=CC(C(C)C)=C1N1[C]N(C=C1)C1=C(C=CC=C1C(C)C)C(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results