missing translation for 'onlineSavingsMsg'
Learn More

Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]silver 97.0+%, TCI America™

Catalog No. C2373200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2373200MG 200 mg
C23731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2373200MG Supplier TCI America Supplier No. C2373200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 873297-19-9
Molecular Formula C27H36AgClN2
Molecular Weight (g/mol) 531.917
InChI Key GNVDTUAJUVJOFQ-UHFFFAOYSA-M
PubChem CID 49788523
IUPAC Name 1,3-bis[2,6-di(propan-2-yl)phenyl]imidazole;chlorosilver
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N2C=CN([C]2)C3=C(C=CC=C3C(C)C)C(C)C.Cl[Ag]

Specifications

Color White
Quantity 200 mg
Formula Weight 531.92
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]silver