accessibility menu, dialog, popup

UserName

CH3H3CONH2

CAS RN 873-95-0

IUPAC Name: 3-amino-5,5-dimethylcyclohex-2-en-1-one
Synonyms: 3-amino-5,5-dimethylcyclohex-2-en-1-one
Molecular Weight (g/mol): 139.2
Molecular Formula: C8H13NO
InChi Key: MTZNODTZOSBYJW-UHFFFAOYSA-N
SMILES: CC1(C)CC(N)=CC(=O)C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results