missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5,5-dimethyl-2-cyclohexen-1-one 98.0+%, TCI America™

Catalog No. A13005G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13005G 5 g
A130025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A13005G Supplier TCI America Supplier No. A13005G

Chemical Identifiers

CAS 873-95-0
Molecular Formula C8H13NO
Molecular Weight (g/mol) 139.198
MDL Number MFCD00045531
InChI Key MTZNODTZOSBYJW-UHFFFAOYSA-N
PubChem CID 136680
IUPAC Name 3-amino-5,5-dimethylcyclohex-2-en-1-one
SMILES CC1(CC(=CC(=O)C1)N)C

Specifications

Melting Point 166°C
Quantity 5 g
Formula Weight 139.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-5,5-dimethyl-2-cyclohexen-1-one