accessibility menu, dialog, popup

UserName

NHOOHCl

CAS RN 87269-87-2

IUPAC Name: benzyl octahydrocyclopenta[b]pyrrole-2-carboxylate hydrochloride
Synonyms: benzyl octahydrocyclopenta[b]pyrrole-2-carboxylate hydrochloride
Molecular Weight (g/mol): 281.78
Molecular Formula: C15H20ClNO2
InChi Key: HLXCXOQXUDRJLF-UHFFFAOYNA-N
SMILES: Cl.O=C(OCC1=CC=CC=C1)C1CC2CCCC2N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results