missing translation for 'onlineSavingsMsg'
Learn More

Benzyl (S,S,S)-2-Azabicyclo[3.3.0]octane-3-carboxylate Hydrochloride 98.0+%, TCI America™

Catalog No. B53445G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B53445G 5 g
B53441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B53445G Supplier TCI America Supplier No. B53445G

Chemical Identifiers

CAS 87269-87-2
Molecular Formula C15H20ClNO2
Molecular Weight (g/mol) 281.78
MDL Number MFCD04974076
InChI Key HLXCXOQXUDRJLF-JKBZPBJLSA-N
Synonym (S,S,S)-2-Azabicyclo[3.3.0]octane-3-carboxylic Acid Benzyl Ester Hydrochloride
PubChem CID 13216857
IUPAC Name benzyl (2S,3aS,6aS)-1,2,3,3a,4,5,6,6a-octahydrocyclopenta[b]pyrrole-2-carboxylate;hydrochloride
SMILES C1CC2CC(NC2C1)C(=O)OCC3=CC=CC=C3.Cl

Specifications

Melting Point 181°C
Color White-Yellow
Quantity 5 g
Formula Weight 281.78
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzyl (S,S,S)-2-Azabicyclo[3.3.0]octane-3-carboxylate Hydrochloride