accessibility menu, dialog, popup

UserName

CAS RN 87-87-6

IUPAC Name: tetrachlorobenzene-1,4-diol
Synonyms: tetrachlorohydroquinone tetrachlorobenzoquinol nordrosophilin A dihydro-p-chloranil usaf DO-62 tetrachloro p-hydroquinone tetrachloro-p-hydroquinone
Molecular Weight (g/mol): 247.88
Molecular Formula: C6H2Cl4O2
InChi Key: STOSPPMGXZPHKP-UHFFFAOYSA-N
SMILES: OC1=C(Cl)C(Cl)=C(O)C(Cl)=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results