missing translation for 'onlineSavingsMsg'
Learn More

Tetrachlorohydroquinone 98.0+%, TCI America™

Catalog No. T00675G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T00675G 5 g
T006725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T00675G Supplier TCI America Supplier No. T00675G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 87-87-6
Molecular Formula C6H2Cl4O2
Molecular Weight (g/mol) 247.88
MDL Number MFCD00041748
InChI Key STOSPPMGXZPHKP-UHFFFAOYSA-N
Synonym tetrachlorohydroquinone, 2,3,5,6-tetrachlorohydroquinone, dihydro-p-chloranil, tetrachloro-p-benzohydroquinone, tetrachlorobenzoquinol, nordrosophilin a, 2,3,5,6-tetrachloro-1,4-benzenediol, perchlorohydroquinone, hydroquinone, tetrachloro, 1,4-benzenediol, 2,3,5,6-tetrachloro
PubChem CID 66603
ChEBI CHEBI:17042
IUPAC Name 2,3,5,6-tetrachlorobenzene-1,4-diol
SMILES C1(=C(C(=C(C(=C1Cl)Cl)O)Cl)Cl)O

Specifications

Melting Point 242°C
Quantity 5 g
Formula Weight 247.88
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetrachlorohydroquinone
RTECSNumber MX7700000
TSCA Yes