accessibility menu, dialog, popup

UserName

CAS RN 87-82-1

IUPAC Name: hexabromobenzene
Synonyms: hexabromobenzene
Molecular Weight (g/mol): 551.49
Molecular Formula: C6Br6
InChi Key: CAYGQBVSOZLICD-UHFFFAOYSA-N
SMILES: BrC1=C(Br)C(Br)=C(Br)C(Br)=C1Br
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)