missing translation for 'onlineSavingsMsg'
Learn More

Hexabromobenzene, 97%

Catalog No. AAB2329522
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2329522 100 g
AAB2329514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2329522 Supplier Thermo Scientific Chemicals Supplier No. B2329522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 87-82-1 | C6Br6 | 551.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87-82-1
Molecular Formula C6Br6
Molecular Weight (g/mol) 551.49
MDL Number MFCD00000058
InChI Key CAYGQBVSOZLICD-UHFFFAOYSA-N
Synonym hexabromobenzene, benzene, hexabromo, perbromobenzene, unii-t01859xwir, ccris 5917, benzene, 1,2,3,4,5,6-hexabromo, hxbbz, hexabioomobenzene, acmc-20aj0o
PubChem CID 6905
IUPAC Name 1,2,3,4,5,6-hexabromobenzene
SMILES C1(=C(C(=C(C(=C1Br)Br)Br)Br)Br)Br

Specifications

Melting Point >300°C
Odor Odorless
Quantity 100 g
Beilstein 1912586
Formula Weight 551.52
Percent Purity 97%
Chemical Name or Material Hexabromobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-773-9
RTECSNumber DA2200100
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only