accessibility menu, dialog, popup

UserName

CAS RN 87-52-5

IUPAC Name: [(1H-indol-3-yl)methyl]dimethylamine
Synonyms: gramin gramine donaxine donaxin gramine (substance)
Molecular Weight (g/mol): 174.25
Molecular Formula: C11H14N2
InChi Key: OCDGBSUVYYVKQZ-UHFFFAOYSA-N
SMILES: CN(C)CC1=CNC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results