missing translation for 'onlineSavingsMsg'
Learn More

Gramine 98.0+%, TCI America™

Catalog No. D065325G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D065325G 25 g
D0653250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D065325G Supplier TCI America Supplier No. D065325G

Chemical Identifiers

CAS 87-52-5
Molecular Formula C11H14N2
Molecular Weight (g/mol) 174.247
MDL Number MFCD00005629
InChI Key OCDGBSUVYYVKQZ-UHFFFAOYSA-N
Synonym gramine, 3-dimethylaminomethyl indole, donaxine, donaxin, gramin, 1h-indole-3-methanamine, n,n-dimethyl, beta-dimethylaminomethylindole, 1-1h-indol-3-yl-n,n-dimethylmethanamine, indol-3-ylmethyldimethylamine, 1h-indol-3-ylmethyl dimethylamine
PubChem CID 6890
ChEBI CHEBI:28948
IUPAC Name 1-(1H-indol-3-yl)-N,N-dimethylmethanamine
SMILES CN(C)CC1=CNC2=CC=CC=C21

Specifications

Melting Point 132°C
Quantity 25 g
Formula Weight 174.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Gramine
RTECSNumber NL7525000
TSCA Yes