accessibility menu, dialog, popup

UserName

CAS RN 869718-80-9

IUPAC Name: 25-azido-2,5,8,11,14,17,20,23-octaoxapentacosane
Synonyms: 25-azido-2,5,8,11,14,17,20,23-octaoxapentacosane
Molecular Weight (g/mol): 409.48
Molecular Formula: C17H35N3O8
InChi Key: ANQOCZRUHGJYCX-UHFFFAOYSA-N
SMILES: COCCOCCOCCOCCOCCOCCOCCOCCN=[N+]=[N-]
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results