missing translation for 'onlineSavingsMsg'
Learn More

25-Azido-2,5,8,11,14,17,20,23-octaoxapentacosane 90.0+%, TCI America™

Catalog No. A272725MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A272725MG 25 mg
A2727100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A272725MG Supplier TCI America Supplier No. A272725MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 869718-80-9
Molecular Formula C17H35N3O8
Molecular Weight (g/mol) 409.48
MDL Number MFCD13184962
InChI Key ANQOCZRUHGJYCX-UHFFFAOYSA-N
PubChem CID 49800635
IUPAC Name 1-azido-2-[2-[2-[2-[2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethane
SMILES COCCOCCOCCOCCOCCOCCOCCOCCN=[N+]=[N-]

Specifications

Color Yellow
Quantity 25 mg
Synonym O-(2-Azidoethyl)-O′C-methylheptaethylene Glycol, mPEG8-Azide, mPEG8-N3, Methyl-PEG8-Azide
Formula Weight 409.48
Percent Purity ≥90.0% (HPLC)
Physical Form Liquid
Chemical Name or Material 25-Azido-2,5,8,11,14,17,20,23-octaoxapentacosane
TSCA No
Recommended Storage Refrigerator