CAS RN 868755-61-7
IUPAC Name:
ethyl 1-methyl-3-(thiophen-2-yl)-1H-pyrazole-5-carboxylate
Synonyms:
ethyl 2-methyl-5-(thiophen-2-yl)pyrazole-3-carboxylate
Molecular Weight (g/mol):
236.29
Molecular Formula:
C11H12N2O2S
InChi Key:
MUIUODSZPZJMMP-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC(=NN1C)C1=CC=CS1
1
–
1
of
1
results
1
Ethyl 1-methyl-3-thien-2-yl-1H-pyrazole-5-carboxylate, 97%, Thermo Scientific™
CAS: 868755-61-7 Molecular Formula: C11H12N2O2S Molecular Weight (g/mol): 236.29 MDL Number: MFCD08060537 InChI Key: MUIUODSZPZJMMP-UHFFFAOYSA-N Synonym: ethyl 1-methyl-3-thien-2-yl-1h-pyrazole-5-carboxylate,ethyl 2-methyl-5-thiophen-2-yl pyrazole-3-carboxylate,ethyl 1-methyl-3-2-thienyl pyrazole-5-carboxylate,ethyl 1-methyl-3-thiophen-2-yl-1h-pyrazole-5-carboxylate,1h-pyrazole-5-carboxylicacid,1-methyl-3-2-thienyl-,ethyl ester,1h-pyrazole-5-carboxylicacid, 1-methyl-3-2-thienyl-, ethyl ester PubChem CID: 7537652 IUPAC Name: ethyl 2-methyl-5-thiophen-2-ylpyrazole-3-carboxylate SMILES: CCOC(=O)C1=CC(=NN1C)C1=CC=CS1