missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-methyl-3-thien-2-yl-1H-pyrazole-5-carboxylate, 97%, Thermo Scientific™

Catalog No. CC47223DA
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC47223DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC47223DA Supplier Thermo Scientific Chemicals Supplier No. CC47223DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 868755-61-7
Molecular Formula C11H12N2O2S
Molecular Weight (g/mol) 236.29
MDL Number MFCD08060537
InChI Key MUIUODSZPZJMMP-UHFFFAOYSA-N
Synonym ethyl 1-methyl-3-thien-2-yl-1h-pyrazole-5-carboxylate, ethyl 2-methyl-5-thiophen-2-yl pyrazole-3-carboxylate, ethyl 1-methyl-3-2-thienyl pyrazole-5-carboxylate, ethyl 1-methyl-3-thiophen-2-yl-1h-pyrazole-5-carboxylate, 1h-pyrazole-5-carboxylicacid,1-methyl-3-2-thienyl-,ethyl ester, 1h-pyrazole-5-carboxylicacid, 1-methyl-3-2-thienyl-, ethyl ester
PubChem CID 7537652
IUPAC Name ethyl 2-methyl-5-thiophen-2-ylpyrazole-3-carboxylate
SMILES CCOC(=O)C1=CC(=NN1C)C1=CC=CS1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1g
Formula Weight 236.29
Percent Purity 97%
Chemical Name or Material Ethyl 1-methyl-3-thien-2-yl-1H-pyrazole-5-carboxylate
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.