accessibility menu, dialog, popup

UserName

CAS RN 868-14-4

IUPAC Name: potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate
Synonyms: potassium bitartrate
Molecular Weight (g/mol): 188.18
Molecular Formula: C4H5KO6
InChi Key: KYKNRZGSIGMXFH-ZVGUSBNCSA-M
SMILES: [K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O
Percent Purity 
  • (3)
  • (14)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
Grade 
  • (4)
  • (6)
  • (4)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (14)
  • (14)
  • (6)
  • (3)
  • (14)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (4)
  • (6)
  • (4)
  • (2)